Skip to Content
Merck
All Photos(1)

Key Documents

929638

Sigma-Aldrich

Poly(ethylene glycol) diacrylamide

Mn 3400, contains MEHQ as stabilizer

Synonym(s):

PEG diacrylamide, PEG hydrogel, PEGDAAm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2CCHC(O)NH(CH2CH2O)n CH2CH2NHC(O)CHCH2
CAS Number:
UNSPSC Code:
12191502
NACRES:
NA.25

form

fine powder

Quality Level

mol wt

Mn 3400
average Mn 3400

contains

≥25 ppm MEHQ as inhibitor
MEHQ as stabilizer

color

white to off-white

solubility

deionized water: 20 wt. % at 23 °C

Ω-end

acrylamide

α-end

acrylamide

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) diacrylamide is a linear, homobifunctional crosslinking PEG polymer with two acrylamide groups. PEG diacrylamide replaces the unstable ester bonds of commonly used PEG diacrylate crosslinkers with hydrolytically stable amides.

Application

PEG diacrylamide is a biocompatible, hydrophilic polymer commonly used to form crosslinked polymer networks and PEG hydrogels. Crosslinked networks can be formed via radical initiator induced polymerization and photopolymerization. Due to its stable amide bond, PEG dacrylamide can be used to prepare hydrogels suitable for long-term studies and eliminate non-specific hydrogel degradation. The acrylamide functional groups can also be reacted with thiol-containing peptide crosslinkers to create enzymatically-sensitive crosslinked networks. PEG diacrylamide has numerous applications in tissue engineering, 3D printing, and drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service