Merck
  • The synthesis and anticancer activity of analogs of the indole phytoalexins brassinin, 1-methoxyspirobrassinol methyl ether and cyclobrassinin.
The synthesis and anticancer activity of analogs of the indole phytoalexins brassinin, 1-methoxyspirobrassinol methyl ether and cyclobrassinin.
Bioorganic & medicinal chemistry (2013-09-10)
Mariana Budovská, Martina Pilátová, Lenka Varinská, Ján Mojžiš, Roman Mezencev
ABSTRACT
An effective synthesis of analogs of the indole phytoalexin cyclobrassinin with NR1R2 group instead of SCH3 was developed starting from indole-3-carboxaldehyde. The target compounds were prepared by spirocyclization of 1-Boc-thioureas with the formation of isolable spiroindoline intermediates, followed by the trifluoroacetic acid-induced cascade reaction consisting of methanol elimination, deprotection and rearrangement of the iminium ion. The structures of novel products were elucided by the (1)H and (13)C NMR spectroscopy, including HMBC, HSQC, COSY, NOESY and DEPT measurements. Several newly synthesized compounds demonstrated significant antiproliferative/cytotoxic activity against human leukemia and solid tumor cell lines, as well as remarkable selectivity of these effects against cancer cells relative to the non-malignant HUVEC cells.
Sigma-Aldrich
Indole-3-carboxaldehyde, 97%