Merck
  • Rab8 and Rabin8-Mediated Tumor Formation by Hyperactivated EGFR Signaling via FGFR Signaling.
Rab8 and Rabin8-Mediated Tumor Formation by Hyperactivated EGFR Signaling via FGFR Signaling.
International journal of molecular sciences (2020-10-24)
Junghwa Choi, Jee Young Sung, Saerom Lee, Jungyoen Yoo, Christopher Rongo, Yong-Nyun Kim, Jaegal Shim
ABSTRACT
The epidermal growth factor receptor (EGFR) signaling is important for normal development, such as vulval development in Caenorhabditis elegans, and hyperactivation of the EGFR is often associated with cancer development. Our previous report demonstrated the multivulva (Muv) phenotype, a tumor model in C. elegans (jgIs25 strain) by engineering LET-23/EGFR with a TKI-resistant human EGFR T790-L858 mutant. Because Rab proteins regulate vesicle transport, which is important for receptor signaling, we screened the RNAi in the jgIs25 strain to find the Rabs critical for Muv formation. Herein, we show that rab-8 RNAi and the rab-8 (-/-) mutation effectively reduce Muv formation. We demonstrate that RABN-8, an ortholog of Rabin8, known as a GEF for Rab8, is also required for Muv formation by promoting the secretion of EGL-17/FGF from vulval precursor cells. In addition, FGFR inhibitors decreased Muv formation mediated by mutant EGFR. Our data suggest that Rab8 and Rabin8 mediate Muv formation through FGF secretion in the EGFR-TKI-resistant nematode model. Furthermore, FGFR-TKIs more effectively inhibit the growth of lung cancer cell lines in H1975 (EGFR T790M-L858R; EGFR-TKI-resistant) than H522 (wild-type EGFR) and H1650 (EGFR exon 19 deletion; EGFR-TKI-sensitive) cells, suggesting that FGFR-TKIs could be used to control cancers with EGFR-TKI-resistant mutations.
Product Description
Millipore
Glutathione−Agarose, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(−)-Tetramisole hydrochloride, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
pGEX-4T-1, Cytiva 28-9545-49