Key Documents
Z377325
Nalgene® bottles, style 2110
size 60 mL
Synonym(s):
nalgene bottle, nalgene bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
polypropylene cap
polypropylene copolymer bottle (PPCO)
square bottle
translucent bottle
wide-mouth bottle
Joint
Joints threaded neck (28)
description
Autoclavable
feature
autoclavable
with cap
packaging
pkg of 12
manufacturer/tradename
Nalgene® 2110-0002
diam. × H
36 mm × 83 mm
size
60 mL
General description
  • Nalgene® square wide mouth bottles are made from polypropylene copolymers.
  • Ideal for storing and transporting various samples.
  • The square design is convenient for labeling.
  • High-quality laboratory-grade materials for low leachable and extractable.
  • Linerless polypropylene closures for sample protection.
  • Autoclavable.
Square, wide-mouth bottles. Leakproof.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)