Skip to Content
Merck
All Photos(1)

Key Documents

SRP6241

Sigma-Aldrich

VAP-1 human

recombinant, expressed in CHO cells, ≥95% (SDS-PAGE)

Synonym(s):

AOC3, Copper Amine Oxidase, HPAO, Membrane Primary Amine Oxidase, SSAO, Vascular Adhesion Protein-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in CHO cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

~80 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VAP1(8639)

General description

VAP-1 (vascular adhesion protein 1) is also called as semicarbazide-sensitive amine oxidase. The VAP-1 gene is mapped to human chromosome 17q21.31.
VAP-1 is a type II membrane cell adhesion protein belonging to the copper/topaquinone oxidase family. It is primarily expressed on the high endothelial venules of peripheral lymph nodes and on hepatic endothelia. VAP-1 can catalyze the oxidative deamination of low molecular weight amines, and plays an important role in the migration of lymphocytes to inflamed tissue. Inhibition of VAP-1 can protect against inflammation related damage to certain injured tissues. Additionally, VAP-1 can function as a significant prognostic marker for certain cancers and cardiovascular diseases. Recombinant VAP-1 is a mixture of monomeric and disulfide linked homodimeric forms of a 737 amino acid polypeptide corresponding to amino acids 27 to 763 of the VAP-1 precursor.

Biochem/physiol Actions

VAP-1 (vascular adhesion protein 1) catalyzes an oxygen-dependent reaction involving the conversion of primary amines to aldehyde, ammonia and hydrogen peroxide. Overexpression of VAP-1 is observed in chronic inflammatory diseases such as chronic renal failure, diabetesand autoimmune disorders. VAP-1 mediates the interaction between leukocytes and endothelium in response to inflammation.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 10 mM Sodium Phosphate, pH 7.8.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Correlation of serum vascular adhesion protein-1 with airflow limitation and quality of life in stable chronic obstructive pulmonary disease
Amani M, et al.
Respiratory Medicine, 132, 149-153 (2017)
Role of vascular adhesion protein-1 in patients with alopecia areata
El-Taweel AEAI, et al.
Beneficial Microbes, 35(1), 28-28 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service