Skip to Content
Merck
All Photos(1)

Key Documents

SML1110

Sigma-Aldrich

Adaphostin

≥98% (HPLC)

Synonym(s):

4-[[(2,5-Dihydroxyphenyl)methyl]amino]benzoic acid tricyclo[3.3.1.13,7]dec-1-yl ester, AG 957 Adamantyl Ester, NSC 680410

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27NO4
CAS Number:
Molecular Weight:
393.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(NCC2=CC(O)=CC=C2O)C=C1)OC34C[C@@]5([H])C[C@@](C[C@@](C5)([H])C4)([H])C3

InChI

1S/C24H27NO4/c26-21-5-6-22(27)19(10-21)14-25-20-3-1-18(2-4-20)23(28)29-24-11-15-7-16(12-24)9-17(8-15)13-24/h1-6,10,15-17,25-27H,7-9,11-14H2/t15-,16+,17-,24-

InChI key

YJZSUCFGHXQWDM-ONHXBKDTSA-N

Biochem/physiol Actions

Adaphostin is a p210bcr/abl tyrosine kinase inhibitor and a potent inducer of myeloid cell death.
Adaphostin is a p210bcr/abl tyrosine kinase inhibitor displaying anti-proliferative activity in leukemia models, human prostate cancer cell line PC-3 and other cancer cell lines. Adaphostin is a potent inducer of myeloid cell death. Similarly to AG957, adaphostin appears to induce a degradation of WT and mutated Bcr/Abl.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koutaroh Okada et al.
EBioMedicine, 41, 105-119 (2019-01-22)
Alectinib has shown a greater efficacy to ALK-rearranged non-small-cell lung cancers in first-line setting; however, most patients relapse due to acquired resistance, such as secondary mutations in ALK including I1171N and G1202R. Although ceritinib or lorlatinib was shown to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service