Skip to Content
Merck
All Photos(1)

Documents

SAB2501711

Sigma-Aldrich

Anti-TFE3 (N-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

RCCP2, TFEA, Transcription factor for IgH enhancer, transcription factor binding to IGHM enhancer 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

bovine, human, mouse, canine

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFE3(7030)

Immunogen

Peptide with sequence SHAAEPARDGVEAS-C, from the N-terminal region of the protein sequence according toNP_006512.2

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service