Skip to Content
Merck
All Photos(1)

Key Documents

HPA006752

Sigma-Aldrich

Anti-USP42 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Deubiquitinating enzyme 42 antibody produced in rabbit, Anti-Ubiquitin carboxyl-terminal hydrolase 42 antibody produced in rabbit, Anti-Ubiquitin thioesterase 42 antibody produced in rabbit, Anti-Ubiquitin-specific-processing protease 42 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PQLPSHMIKNPPHLNGTGPLKDTPSSSMSSPNGNSSVNRASPVNASASVQNWSVNRSSVIPEHPKKQKITISIHNKLPVRQCQSQPNLHSNSLENPTKPVPSSTITNSAVQSTSNASTMSVSSKVTK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP42(84132)

Immunogen

Ubiquitin carboxyl-terminal hydrolase 42 recombinant protein epitope signature tag (PrEST)

Application

Anti-USP42 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

USP42 (Ubiquitin specific peptidase 42) is a nuclear protein belonging to the ubiquitin-specific protease (USP) family. It is widely distributed in human tissues, especially in skeletal muscle and testis. It is involved in the regulation of ubiquitin-mediated pathways. It exerts deubiquitylating activity by cleaving ubiquitin from an ubiquitin-β-galactosidase fusion protein. It plays an important role in the regulation of p53 (a tumor suppressor) activity. It helps to stabilize p53 in stress response and promotes early activation of p53.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70793

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Spolverini et al.
Oncogene, 36(42), 5819-5828 (2017-06-13)
Monoubiquitylation of histone H2B (H2Bub1) is catalyzed mainly by the RNF20/RNF40 complex and erased by multiple deubiquitylating enzymes (DUBs). H2Bub1 influences many aspects of chromatin function, including transcription regulation and DNA repair. Cancer cells often display reduced levels of H2Bub1
Andreas K Hock et al.
The EMBO journal, 30(24), 4921-4930 (2011-11-17)
The p53 tumour suppressor protein is a transcription factor that prevents oncogenic progression by activating the expression of apoptosis and cell-cycle arrest genes in stressed cells. The stability of p53 is tightly regulated by ubiquitin-dependent degradation, driven mainly by the
Nagore Elu et al.
Frontiers in physiology, 10, 534-534 (2019-05-28)
The ubiquitin E3 ligase UBE3A has been widely reported to interact with the proteasome, but it is still unclear how this enzyme regulates by ubiquitination the different proteasomal subunits. The proteasome receptor DDI1 has been identified both in Drosophila photoreceptor
Nicole Giebel et al.
EMBO reports, 22(5), e51415-e51415 (2021-04-01)
The tumour suppressors RNF43 and ZNRF3 play a central role in development and tissue homeostasis by promoting the turnover of the Wnt receptors LRP6 and Frizzled (FZD). The stem cell growth factor R-spondin induces auto-ubiquitination and membrane clearance of ZNRF3/RNF43
Víctor Quesada et al.
Biochemical and biophysical research communications, 314(1), 54-62 (2004-01-13)
We have identified and cloned 22 human cDNAs encoding novel members of the ubiquitin-specific protease (USP) family. Eighteen of the identified proteins contain all structural features characteristic of these cysteine proteinases, whereas four of them have been classified as non-peptidase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service