Skip to Content
Merck
All Photos(1)

Key Documents

L6.G8

92102117, rat muscle, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

L6.G8, 92102117, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 92102117, rat muscle, Fibroblast

biological source

rat muscle

packaging

tube of 5 μg 92102117-DNA-5UG
pkg of vial of cells 92102117-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat skeletal muscle myoblast

Cell Line Description

A subclone of L6, a rat thigh muscle cell line. Previously, on reaching confluence the cells fused to form myotubes and a twitching movement could be observed. However, these cells have lost the ability to fuse after reaching confluency.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures to a seeding density of 1.5-2x1,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37 C. Cells have lost their ability to fuse after reaching confluency.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service