Skip to Content
Merck
All Photos(1)

Key Documents

L35

91012319, Porcine blood, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

L35, 91012319

biological source

Porcine blood

packaging

tube of 5 μg 91012319-DNA-5UG
pkg of vial of cells 91012319-1VL

growth mode

Suspension

karyotype

Modal no. 36.5

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Porcine peripheral blood T cell

Cell Line Description

One of a series of cell lines established from mononuclear cells of a boar (Sus scrofa domestica L) which was injected with Shimozuma cells producing porcine retrovirus (Tsukuba-1). The cells express MSA4. The cells contain retrovirus particles and are positive in RT assay.

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 1 mM Sodium Pyruvate (NaP) + 0.05 mM 2-Mercaptoethanol (2ME) + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 100,000 to 1,000,000 cells/ml; 5% CO2; 37 °C. Cells grow as small clumps.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service