Skip to Content
Merck
All Photos(1)

Key Documents

C8166

88051601, human blood, Lymphoblast

Synonym(s):

C8166-CD4 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

C8166, 88051601

biological source

human blood

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Lymphoblast

products

Human T Lymphotrohpic Virus 1 (HTLV-1) tat gene

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human T cell leukaemia

Cell Line Description

Clone of C63/CRII-4 derived by fusion of primary umbilical cord blood cells with HTLV-1 producing line from adult T cell leukaemia lymphoma patient. Contain defective HTLV-1 genome. In presence of HIV cells fuse to form syncytia. The cells should be handled under laboratory containment level 2.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells grow in aggregates.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service