Skip to Content
Merck

02192

UV-VIS detectable IEF Marker

BioChemika, pI 7.7

Synonym(s):

IEF Marker, UV-VIS detectable

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352201
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioChemika

storage temp.

2-8°C

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Still not finding the right product?

Explore all of our products under UV-VIS detectable IEF Marker


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library