Skip to Content
Merck
All Photos(1)

Key Documents

S26-M

Sigma-Aldrich

Goat Serum, 1 L

Non-sterile goat serum collected from normal healthy goats. Used for blocking of interfering antibodies.

Synonym(s):

1L Goat Serum, Goat Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105901
eCl@ss:
26020100
NACRES:
NA.71
biological source:
goat
Pricing and availability is not currently available.

biological source

goat

antibody form

serum

manufacturer/tradename

Chemicon®

technique(s)

blocking: suitable (interfering antibodies)

General description

Normal goat serum from healthy animals collected from USDA inspected facilities. All animals have received ante and post mortem inspections and were found to be free of contagious diseases.

HEMOGLOBIN: ~3.5 mg/dL

COUNTRY OF ORIGIN: USA

Application

Blocking reagent. Dilute as needed with PBS or other buffer. Typical working concentrations are 0.1-5% serum in buffer. For best results, it is recommended to filter the diluted serum solution through a 0.2-0.45μm membrane prior to use.Blocking reagent; dilute as needed with PBS or other buffer. Typical working concentrations are 0.1-5% serum in buffer.

Optimal dilutions determined by enduser.

Physical form

Liquid clean serum, unfiltered, containing no preservatives.

Storage and Stability

Store frozen at -20°C in undiluted aliquots for up to twelve months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Is it normal for lot 3671514 to have a more amber or orangeish color compared to previous lots?

    1 answer
    1. Yes, the more amber or orangeish color in lot 3671514 is due to the presence of hemoglobin, which is normal for batches with higher hemoglobin content. This change in color is consistent with serum products regardless of species and age of donor.

      Helpful?

  2. What's the difference between S26-100ML and S26-100KC?

    1 answer
    1. Product S26 is available in 100ml (S26-100ML) and 1L (S26-LITER) formats. Regarding the S26-100KC code, this is on the product page as an option for bulk or large quantity orders. It is intended for quantities in excess of 10-20 times the largest format available in the catalog.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service