Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
7F9, monoclonal
Application:
IF, IHC, IP, WB
Citations:
-
biological source
mouse
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
7F9, monoclonal
species reactivity
rat
packaging
antibody small pack of 25 μg
technique(s)
immunofluorescence: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
rat ... Cpm(314855)
General description
50.32 kDa calculated.
Carboxypeptidase M (UniProt: D4A9Q5; also known as CPM) is encoded by the Cpm gene (Gene ID: 314855) in rat. Rat CPM a glycosylphosphatidylinositol (GPI) anchored metallo-carboxypeptidase that cleaves basic carboxyl-terminal amino acids from various peptides at neutral pH. Rat CPM is synthesized with a signal peptide (aa 1-17), which is subsequently cleaved off to generate the mature form. The mature CPM displays high specificity and cleaves only C-terminal arginine or lysine residues in a variety of peptide substrates. It is shown to be expressed in the lung and placenta at high levels. Its expression is detected in alveolar cells lacking T1alpha, a type I cell marker protein found in adult lung. Carboxypeptidase M also serves as a macrophage differentiation marker, which is preferentially induced in epithelioid cells of all granuloma types studied, but not in resting macrophages. Hence, it can be employed for detection of minute granuloma even in the presence of non-granulomatous macrophages. Clone 7F9 recognizes CPM expressed in late embryonic rat lung to postnatal day 1. (Ref.: Fujiwara, N., et al. (2007), Respirology 12(1); 54-62; Tsakiris, I., et al. (2012). Lab. Invest. 92(3); 345-361).
Immunogen
Epitope: extracellular domain
Membrane fraction of E18.5 embryonic rat lung.
Application
Anti-carboxypeptidase M, clone 7F9, Cat. No. MABS1925, is a mouse monoclonal antibody that detects Carboxypeptidase M and has been tested for use in Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Signaling
Signaling
Western Blotting Analysis: A representative lot detected carboxypeptidase M in Western Blotting applications (Fujiwara, N., et. al. (2007). Respirology. 12(1):54-62).
Immunoprecipitation Analysis: A representative lot immunoprecipitated carboxypeptidase M in Immunoprecipitation applications (Fujiwara, N., et. al. (2007). Respirology. 12(1):54-62).
Immunohistochemistry Analysis: A representative lot detected carboxypeptidase M in Immunohistochemistry applications (Fujiwara, N., et. al. (2007). Respirology. 12(1):54-62).
Immunoprecipitation Analysis: A representative lot immunoprecipitated carboxypeptidase M in Immunoprecipitation applications (Fujiwara, N., et. al. (2007). Respirology. 12(1):54-62).
Immunohistochemistry Analysis: A representative lot detected carboxypeptidase M in Immunohistochemistry applications (Fujiwara, N., et. al. (2007). Respirology. 12(1):54-62).
Biochem/physiol Actions
Clone 7F9 specifically detects carboxypeptidase M in rat tissues.
Physical form
Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Immunofluorescence in E19 rat embryo tissue sections.
Immunofluorescence Analysis: A 1:25 dilution of this antibody detected carboxypeptidase M in E19 rat embryo tissue sections.
Immunofluorescence Analysis: A 1:25 dilution of this antibody detected carboxypeptidase M in E19 rat embryo tissue sections.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.