Skip to Content
Merck
All Photos(1)

Key Documents

MABF839

Sigma-Aldrich

Anti-Ly-6G Antibody, clone 1A8 (Azide free)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

Quality Level

clone

1A8
monoclonal

purified by

using protein G

species reactivity

mouse, rat

concentration

(Please refer to lot specific datasheet.)

technique(s)

activity assay: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

UniProt accession no.

General description

Propeitary whole cells expressing to mouse Ly-6G.

Application

Flow Cytometry Analysis: 0.5 µg of this antibody from a representative lot detected Ly-6G in 1X10E6 mouse bone marrow cells.
Flow Cytometry Analysis: A representative lot detected Ly-6G in transfected EL4 cells and bone marrow cells (Fleming, T.J. et al. (1993). Journal of Immunology. 151(5):2399-2408).
Activity Assay Analysis: A representative lot detected Ly-6G in transmission of S. pneumoniae from index after injection with this clone (Short, K.R., et al. (2013). mBio. 3(5):e00255-12).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Ly-6G in transfected EL4 cells (Fleming, T.J. et al. (1993). Journal of Immunology. 151(5):2399-2408).

Quality

Evaluated by Flow Cytometry in mouse splenocyte.

Flow Cytometry Analysis: 0.5 µg of this antibody detected Ly-6G in mouse splenocyte.

Physical properties

12 kDa calculated

Physical form

Purified rat monoclonal IgG2aκ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service