Skip to Content
Merck
All Photos(1)

Key Documents

556746

Sigma-Aldrich

RNase A, Protease-Free, Highly Purified, Bovine Pancreas

For selective degradation of RNA

Synonym(s):

RNase A, Protease-Free, Highly Purified, Bovine Pancreas, Ribonuclease A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

Quality Level

description

Merck USA index - 14, 8202

Assay

≥95% (SDS-PAGE)

form

lyophilized solid

specific activity

81-99 units/mg material

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

water: 2 mg/mL

foreign activity

Protease, none detected

shipped in

ambient

storage temp.

−20°C

SMILES string

[nH]1cnc(c1)CC(NC(=O)CCN)C(=O)O

InChI

1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)

InChI key

CQOVPNPJLQNMDC-UHFFFAOYSA-N

General description

Note: 1 KU = 1000 units.
Ribonuclease A, Protease-Free is a

chromatographically purified, pyrimidine-specific

endoribonuclease that acts on single-stranded

RNA.
Ribonuclease A, Protease-Free is achromatographically purified, pyrimidine-specificendoribonuclease that acts on single-strandedRNA.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the hydrolysis of RNA to yield a first-order velocity constant equal to 1 at 25°C, pH 5.0.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Analysis Note

May contain trace amounts of DNase I, which can be inactivated by boiling for 15 min in 50 mM potassium acetate, pH 5.5.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service