Skip to Content
Merck
All Photos(1)

Documents

5.04043

Sigma-Aldrich

Rho Inhibitor II, Y16

Synonym(s):

Rho Inhibitor II, Y16, PDZ Rho-Binding Domain Blocker, p115 Rho-Binding Domain Blocker, LARG Rho-Binding Domain Blocker, (E)-4-(3-(3-Methylbenzyloxy)benzylidene)-1-phenylpyrazolidine-3,5-dione, (4E)-4-(3-(3-Methylbenzyloxy)benzylidene)-1-phenyl-3,5-pyrazolidinedione, p115 Rho-Binding Domain Blocker, PDZ Rho-Binding Domain Blocker, LARG Rho-Binding Domain Blocker, (E)-4-(3-(3-Methylbenzyloxy)benzylidene)-1-phenylpyrazolidine-3,5-dione, (4E)-4-(3-(3-Methylbenzyloxy)benzylidene)-1-phenyl-3,5-pyrazolidinedio

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20N2O3
CAS Number:
Molecular Weight:
384.43
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

SMILES string

CC1=CC(=CC=C1)COC2=CC=CC(=C2)C=C3C(=O)NN(C3=O)C4=CC=CC=C4

General description

A cell-permeable pyrazolidinedione compound that is shown to target RhoGEF DH-PH domain junction with high affinity (Kd = 65 nM) and effetively prevent RhoGEFs LARG, p115, and PDZ from interacting with RhoA, while displaying little potency against DBL-RhoA, LBC-RhoA, intersectin-Cdc42, or TrioN-Cdc42 interaction. Shown to completely prevent serum-induced activation of cellular RhoA, but not Cdc42 or Rac1, in NIH-3T3 cultures (10 µM) and RhoA downstream signaling events. Greatly synergizes with Rho GEF-binding domain blocker Rhosin (Cat. No. 555460) in blocking RhoA-LARG interaction and in preventing cellular RhoA activation (both drug at 5 µM).
A cell-permeable pyrazolidinedione compound that prevents RhoGEFs LARG, p115, and PDZ from interacting with RhoA, while displaying little potency against DBL-RhoA, LBC-RhoA, intersectin-Cdc42, or TrioN-Cdc42 interaction. Structure and in vitro binding studies using DH-PH domains-containing human LARG catalytic fragment (aa 765-1138) constructs reveals RhoGEF DH-PH domain junction as the high affinity Y16 targeting site (Kd = 65, 472, 2121, and >5 x 105 nM using wt, K979A, E982A, and N983A LARG, respectively). Shown to prevent serum-induced activation of cellular RhoA, but not Cdc42 or Rac1, in serum-starved NIH-3T3 cultures (215%, 109%, and 87% of basal RhoA-GTP level with 0, 10, or 30 µM Y16, respectively) and effectively inhibit RhoA downstream signaling events, including FAK and MLC phosphorylations and stress fiber formation without apparent cytotoxicity (50 µM up to 24 h) or affecting Cdc42-mediated filapodia and Rac1-mediated lamellipodia formation. Greatly synergizes with Rho GEF-binding domain blocker Rhosin (Cat. No. 555460) in blocking RhoA-LARG binding (8%, 27%, and 72% inhibition, respectively, with 5 µM G04 alone, 5 µM Y16 alone, or in combination) and in inhibiting cellular RhoA activation (80% inhibition of serum-stimulated RhoA-GTP level by 30 µM Y16 alone or by a combination of 5 µM each of G04 and Y16 in serum-starved NIH-3T3 cells).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RhoGEF
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze -20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shang, X., et al. 2013. Proc. Natl. Acad. Sci. USA.110, 3155.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service