Skip to Content
Merck
All Photos(1)

Key Documents

166900

Millipore

Fluid A

bottle of, ready-to-use, bottle volume 100 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type screw cap with septum
ready-to-use

packaging

bottle of

technique(s)

sterile filtration: suitable

bottle capacity

125 mL

bottle volume

100 mL , filling volume

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Fluid A is a general rinse buffer designed in compliancewith the European Pharmacopeia (EP) 2.6.1, United States Pharmacopeia (USP) 71and Japanese Pharmacopeia (JP) 4.06. The product is a clear colorless liquid.

Each bottle has a screw cap with septum closure to reducethe risk of cross-contamination and optimize the disinfection procedures.

Application

Fluid A is used to rinse aqueous solutions during sterilitytesting by membrane filtration. It is also used to dilute soluble solids forthe same application and as a rinsing fluid for membrane filtration ofnon-sterile products.
The product is robust and suitable for:
  • Dissolving or diluting samples,
  • Reconstituting commercial microorganism sample.
  • As a transport medium for microorganisms

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service