1.07536
QUARTZ FINE GRANULAR,
WASHED AND CALCINED FOR ANALYSIS
Synonym(s):
QUARTZ FINE GRANULAR,
About This Item
Quality Level
form
solid
impurities
≤0.05% HCl solubles
loss
≤0.05% loss on ignition, 900 °C
particle size
0.2-0.8 mm (>=40%)
pH
5-8 (20 °C, 400 g/L in H2O)
bp
>2200 °C/1013 hPa
mp
1713 °C
density
2.65 g/cm3 at 20 °C
bulk density
1400 kg/m3
anion traces
chloride (Cl-): ≤0.01%
storage temp.
no temp limit
SMILES string
[Si](=O)=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Analysis Note
Chloride (Cl): ≤ 0.01 %
Loss on ignition (900 °C): ≤ 0.05 %
Particle size (0.2 - 0.8 mm): ≥ 40 %
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service