Skip to Content
Merck
All Photos(1)

Key Documents

860616P

Avanti

Sphingosyl PI (d18:1)

Avanti Research - A Croda Brand 860616P, powder

Synonym(s):

D-erythro-sphingosyl phosphoinositol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H48NO10P
CAS Number:
Molecular Weight:
541.61
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860616P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860616P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])([NH3+])COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O

InChI

1S/C24H48NO10P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(26)17(25)16-34-36(32,33)35-24-22(30)20(28)19(27)21(29)23(24)31/h14-15,17-24,26-31H,2-13,16,25H2,1H3,(H,32,33)/b15-14+/t17-,18+,19?,20+,21?,22?,23?,24+/m0/s1

InChI key

NFRGTPKVKNBLRN-VBGCAUIASA-N

Application

Sphingosyl PI (d18:1) has been used in the synthesis of D-erythro-N-palmitoyl-inositol-phosphorylceramide.

Packaging

5 mL Amber Glass Screw Cap Vial (860616P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of membrane properties of inositol phosphorylceramide
Bjorkbom A, et al.
Biochimica et Biophysica Acta - Biomembranes, 1798(3), 453-460 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service