Z503800
Scienceware® trays
w/ faucet, inside dimensions L × W × H 30 cm × 41 cm × 7.6 cm
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
Recommended Products
material
low-density polyethylene container
manufacturer/tradename
Bel-Art
Related Categories
General description
Molded LDPE can withstand temperatures up to 77°C (171°F). The PE faucet has a 12.7 mm bore.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)