Key Documents
Z261149
Sigma-Aldrich
Nalgene® PassPort IP2 bottles
Wide-mouth, capacity 250 mL
Synonym(s):
nalgene bottle, nalgene bottles, nalgene hdpe
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
high-density polyethylene
polypropylene cap
round translucent bottle
Joint
Joints threaded neck (43-415)
feature
with cap
packaging
pack of 12 (bottles)
manufacturer/tradename
Nalgene® 2199-0008
bottle diam. × H
61 mm × 131 mm
capacity
250 mL
General description
Bottles suitable for shipping. These bottles are in compliance with the new United Nations Performance-Oriented Packaging regulations for shipping. IP2 marking, material, volume, and registration notch molded into bottom of bottle. Leakproof. Evaluated at 15 psi.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
PassPort is a trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)