Key Documents
Z183598
Sigma-Aldrich
Cryo-apron®
size 24 in. × 36 in.
About This Item
UNSPSC Code:
46181501
NACRES:
NB.35
manufacturer/tradename
TempShield CA36
size
24 in. × 36 in.
General description
Provides protection in extreme environments. Resistant to solvents/chemicals. Breathable, static-free, and asbestos-free. 100% waterproof. Reversible. Adjustable straps at neck and waist.
Legal Information
Cryo-apron is a registered trademark of Tempshield, Inc.