Skip to Content
Merck
All Photos(1)

Key Documents

GF58380318

Polymethylmethacrylate

(PMMA, ACRYLIC), sphere, diam. 1.5 mm, weight 50 pcs, tolerance ±0.05mm

Synonym(s):

ME306810

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

form

spheres

manufacturer/tradename

Goodfellow 583-803-18

diam.

1.5 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service