Skip to Content
Merck
All Photos(1)

Documents

GF02721710

Polycarbonate

PC - 30% Carbon Fiber Reinforced, granule, 3 mm nominal granule size, weight 200 g, black

Synonym(s):

CT356310

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

product name

Polycarbonate - 30% Carbon Fiber Reinforced, (PC - 30% CFR), granule, 3 mm nominal granule size, weight 200 g, black

form

granular

manufacturer/tradename

Goodfellow 027-217-10

weight

200 g

color

black

nominal granule size

3 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service