DWK213725479
DURAN® Volumetric flask partially ground without print
neck joint: ST/NS 24/29, flask capacity (1,000 mL)
Synonym(s):
213725479
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Joint
joint: ST/NS neck24/29
feature
class A with certificate
packaging
case of 10 ea
manufacturer/tradename
DURAN® 213725479
flask capacity
(1,000 mL)
fitting
thread (24/29)
Looking for similar products? Visit Product Comparison Guide
General description
Calibration based on the poured in volume (“In”) at a + 20 °C reference temperature. Thanks to the exact processing and precisely calibrated scales, they permit the highly accurate determination and measurements of volume. Supplied with ergonomic polyethylene stopper for easy and safe tightening or removal. The volume content tolerances conform to accuracy Class A with batch certificate and certificate of conformity. Fired-on, highly durable blue print. With printed ring mark for better visibility. Influence of undesirable factors is kept to an absolute minimum
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service