CLS5600500
Pyrex® Lifetime-Red™ volumetric flask with snap cap
capacity 500 mL
Synonym(s):
clear flasks, pyrex flask, volumetric flasks, Corning® 5600
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
long
sterility
non-sterile
feature
class A
closure type Snap cap
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 5600-500
tol. measuring range
0.20 mL
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
General description
Capacity mark is fine line-etched through narrow, red band into clear glass.
Legal Information
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service