CLS560050
Pyrex® Lifetime-Red™ volumetric flask with snap cap
capacity 50 mL
Synonym(s):
clear flasks, pyrex flask, volumetric flasks, Corning® 5600
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
long
sterility
non-sterile
feature
closure type Snap cap
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 5600-50
tol. measuring range
0.05 mL
capacity
50 mL
General description
Capacity mark is fine line-etched through narrow, red band into clear glass.
Legal Information
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.