Key Documents
BAH134160001
Dial calipers
L 222 mm
About This Item
UNSPSC Code:
41111700
NACRES:
NB.79
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H13416-0001
L
222 mm
Related Categories
General description
Precision plastic calipers with Metric and English scales with 0.1 mm and 0.001 in. subdivisions. Use the thumb roller for precise measurements. Parallel jaws are 40 mm (1 ½ in.) deep.
  • Non scratching, non-conductive and corrosion resistant
  • Inside and outside jaws and round plastic rod depth gauge allow most measurements
Choose from one of the most recent versions:
Certificates of Analysis (COA)