588253
Biuret-13C2
99 atom % 13C, 97% (CP)
Synonym(s):
Imidodicarbonic-13C2 diamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
isotopic purity
99 atom % 13C
Assay
97% (CP)
form
solid
bp
185-190 °C (dec.)(lit.)
mass shift
M+2
SMILES string
N[13C](=O)N[13C](N)=O
InChI
1S/C2H5N3O2/c3-1(6)5-2(4)7/h(H5,3,4,5,6,7)/i1+1,2+1
InChI key
OHJMTUPIZMNBFR-ZDOIIHCHSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service