485764
Benzyl-2,3,4,5,6-d5 chloride
98 atom % D
Synonym(s):
α-Chlorotoluene-2,3,4,5,6-d5
About This Item
Recommended Products
isotopic purity
98 atom % D
Quality Level
Assay
99% (CP)
refractive index
n20/D 1.538 (lit.)
bp
177-181 °C (lit.)
mp
−43 °C (lit.)
density
1.144 g/mL at 25 °C
mass shift
M+5
SMILES string
[2H]c1c([2H])c([2H])c(CCl)c([2H])c1[2H]
InChI
1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2/i1D,2D,3D,4D,5D
InChI key
KCXMKQUNVWSEMD-RALIUCGRSA-N
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3
Target Organs
Heart,forestomach, Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
140.0 °F - closed cup
Flash Point(C)
60.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service