Skip to Content
Merck
All Photos(1)

Key Documents

275425

Sigma-Aldrich

2,5-Dimethoxythiophenol

98%

Synonym(s):

2,5-Dimethoxybenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3SH
CAS Number:
Molecular Weight:
170.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

refractive index

n20/D 1.5860 (lit.)

bp

88-90 °C/0.5 mmHg (lit.)

density

1.134 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(OC)c(S)c1

InChI

1S/C8H10O2S/c1-9-6-3-4-7(10-2)8(11)5-6/h3-5,11H,1-2H3

InChI key

SESUUAOAUZDHHP-UHFFFAOYSA-N

General description

The s-alkylation of 2,5-dimethoxythiophenol with pentyl bromide and heptyl bromide was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Open-ring models of hetero-cannabinoids: synthesis of 2-(alkylthio)-5-(2-aminoethyl) hydroquinone derivatives.
Collection of Czechoslovak Chemical Communications, 55(3), 782- 796 (1990)
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service