跳转至内容
Merck
  • Prevention of water vapour adsorption by carbon molecular sieves in sampling humid gases.
Prevention of water vapour adsorption by carbon molecular sieves in sampling humid gases.
Journal of chromatography. A (2002-01-05)
M Gawryś, P Fastyn, J Gawlowski, T Gierczak, J Niedzielski
摘要
The water uptake by the solid sorbents Carbosieve S-III, Carboxen 569, 1000 and 1001, all of which are used for sampling of volatile organic compounds from the atmosphere. was examined using a direct experimental approach. The content of retained water is affected both by the trap temperature and the initial water vapour concentration in the sampled gas. Two different adsorption mechanism are operative. At low relative humidities (RH) only active polar centres are involved. This adsorption is so weak that negative water interferences can easily be managed. Another mechanism, the micropore volume filling, involves substantial amounts of water, becomes operative once the threshold value for relative humidity (RHth) is surpassed. RHth is 45+/-3% for Carboxen 1000 decreasing to 35+/-3% for the three other sorbents studied. A novel but simple strategy was tested for water management: moderate heating of the trap during the sampling (a warm trap method). The temperature elevation required depends on the RHth characteristic for the specific sorbent, and RH and the temperature of the sampled gas. Usually the 5-15 degrees C elevation is sufficient; only under extreme RH conditions is an elevation of 20 degrees C necessary. The diagrams are given to determine this elevation. Since the sample RH is significantly decreased at an elevated temperature the negative effect of water uptake on the safe sampling volume is alleviated. Consequently the sampled gas volume can be as large as desired which decreases detection limits.
材料
货号
品牌
产品描述
Supelco
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Supelco
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Supelco
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
ORBO Carboxen-564 管, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO-78 400/200mg,每包 25 支, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
Carboxen®1010 多孔层壁涂开管柱, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
SPME固相微萃取萃取头 Carboxen/聚二甲基硅氧烷 (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
Carbopack®吸附剂, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen® 1006 多孔层壁涂开管柱, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
SPME固相微萃取萃取头 Carboxen/聚二甲基硅氧烷 (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME固相微萃取萃取头 Carboxen/聚二甲基硅氧烷 (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME固相微萃取萃取头 Carboxen/聚二甲基硅氧烷 (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
Carboxen®1010 多孔层壁涂开管柱, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carbosieve 吸附剂, matrix Carbosieve S-III, 60-80 mesh, bottle of 10 g
Supelco
SPME固相微萃取萃取头 Carboxen/聚二甲基硅氧烷 (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME固相微萃取萃取头 Carboxen/聚二甲基硅氧烷 (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME固相微萃取萃取头 Carboxen/聚二甲基硅氧烷 (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Supelco
Carboxen® 1006 多孔层壁涂开管柱, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® 1000 双向固相萃取管, 200 mg/mL, pk 30
Supelco
Carbopack®吸附剂, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Supelco
Carbopack®吸附剂, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Supelco
Tenax® TA/Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
Carbopack®吸附剂, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
Carbopack®吸附剂, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
ORBO-93吸附管, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Supelco
Carbosieve 吸附剂, matrix Carbosieve S-II, 60-80 mesh, bottle of 10 g
Supelco
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm