跳转至内容
Merck
  • Recessive truncating titin gene, TTN, mutations presenting as centronuclear myopathy.
Recessive truncating titin gene, TTN, mutations presenting as centronuclear myopathy.
Neurology (2013-08-27)
Ozge Ceyhan-Birsoy, Pankaj B Agrawal, Carlos Hidalgo, Klaus Schmitz-Abe, Elizabeth T DeChene, Lindsay C Swanson, Rachel Soemedi, Nasim Vasli, Susan T Iannaccone, Perry B Shieh, Natasha Shur, Jane M Dennison, Michael W Lawlor, Jocelyn Laporte, Kyriacos Markianos, William G Fairbrother, Henk Granzier, Alan H Beggs
摘要
To identify causative genes for centronuclear myopathies (CNM), a heterogeneous group of rare inherited muscle disorders that often present in infancy or early life with weakness and hypotonia, using next-generation sequencing of whole exomes and genomes. Whole-exome or -genome sequencing was performed in a cohort of 29 unrelated patients with clinicopathologic diagnoses of CNM or related myopathy depleted for cases with mutations of MTM1, DNM2, and BIN1. Immunofluorescence analyses on muscle biopsies, splicing assays, and gel electrophoresis of patient muscle proteins were performed to determine the molecular consequences of mutations of interest. Autosomal recessive compound heterozygous truncating mutations of the titin gene, TTN, were identified in 5 individuals. Biochemical analyses demonstrated increased titin degradation and truncated titin proteins in patient muscles, establishing the impact of the mutations. Our study identifies truncating TTN mutations as a cause of congenital myopathy that is reported as CNM. Unlike the classic CNM genes that are all involved in excitation-contraction coupling at the triad, TTN encodes the giant sarcomeric protein titin, which forms a myofibrillar backbone for the components of the contractile machinery. This study expands the phenotypic spectrum associated with TTN mutations and indicates that TTN mutation analysis should be considered in cases of possible CNM without mutations in the classic CNM genes.
材料
货号
品牌
产品描述
Sigma-Aldrich
Monoclonal Anti-Ryanodine Receptor antibody produced in mouse, clone C3-33, ascites fluid
Sigma-Aldrich
Monoclonal Anti-TTN antibody produced in mouse, clone 7D3, purified immunoglobulin, buffered aqueous solution