跳转至内容
Merck
Mammalian Sterile20-like kinase 1 and the regulation of apoptosis.
Biochemical Society transactions (2004-05-26)
P M de Souza, M A Lindsay
摘要
Mammalian Sterile20-like kinase 1 (Mst1) is a ubiquitously expressed serine/threonine kinase which represents a member of the rapidly expanding family of enzymes related to the yeast Sterile20 kinase. Although the physiological function of Mst1 and its role in intracellular signalling is still unclear, reports to date suggest that Mst1, similar to its yeast homologue, operates in the MAPK (mitogen-activated protein kinase) pathway and, in this capacity, may represent a putative MAPK kinase kinase kinase. Moreover, there is abundant evidence for a role of this enzyme in apoptosis, where not only is it a target for caspases, but may also serve as an activator of these proteases to amplify the apoptotic signalling pathway. This paper reviews the investigations that have led to our current understanding of the mechanisms by which Mst1 may be activated and thereby contribute to apoptosis.
材料
货号
品牌
产品描述
Sigma-Aldrich
MST1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution