跳转至内容
Merck
  • Bacterial Lipopolysaccharides Induce Steroid Sulfatase Expression and Cell Migration through IL-6 Pathway in Human Prostate Cancer Cells.
Bacterial Lipopolysaccharides Induce Steroid Sulfatase Expression and Cell Migration through IL-6 Pathway in Human Prostate Cancer Cells.
Biomolecules & therapeutics (2013-09-07)
Hee-Jung Im, Na-Hee Park, Yeo-Jung Kwon, Sangyun Shin, Donghak Kim, Young-Jin Chun
摘要
Steroid sulfatase (STS) is responsiblefor the conversion of estrone sulfate to estrone that can stimulate growth in endocrine-dependent tumors such as prostate cancer. Although STS is considered as a therapeutic target for the estrogen-dependent diseases, cellular function of STS are still not clear. Previously, we found that tumor necrosis factor (TNF)-α significantly enhances steroid sulfatase expression in PC-3 human prostate cancer cells through PI3K/Akt-dependent pathways. Here, we studied whether bacterial lipopolysaccharides (LPS) which are known to induce TNF-α may increase STS expression. Treatment with LPS in PC-3 cells induced STS mRNA and protein in concentration- and time-dependent manners. Using luciferase reporter assay, we found that LPS enhanced STS promoter activity. Moreover, STS expression induced by LPS increased PC-3 tumor cell migration determined by wound healing assay. We investigated that LPS induced IL-6 expression and IL-6 increased STS expression. Taken together, these data strongly suggest that LPS induces STS expression through IL-6 pathway in human prostate cancer cells.
材料
货号
品牌
产品描述
Sigma-Aldrich
MISSION® esiRNA, targeting human STS