跳转至内容
Merck
  • Targeting the Neurokinin-1 Receptor Compromises Canonical Wnt Signaling in Hepatoblastoma.
Targeting the Neurokinin-1 Receptor Compromises Canonical Wnt Signaling in Hepatoblastoma.
Molecular cancer therapeutics (2015-10-31)
Matthias Ilmer, Agnès Garnier, Jody Vykoukal, Eckhard Alt, Dietrich von Schweinitz, Roland Kappler, Michael Berger
摘要
The substance P (SP)/NK-1 receptor (NK1R) complex represents an intriguing anticancer target for a variety of tumors, including hepatoblastoma (HB). Therefore, NK1R antagonists, such as the clinical drug aprepitant, recently have been proposed as potent anticancer agents. However, very little is known regarding the molecular basis of NK1R inhibition in cancer. Using reverse phase protein array, Western blot, Super TOP/FOP, confocal microscopy, and sphere formation ability (SFA) assays, we identified the AKT and Wnt signaling pathways as the key targets of aprepitant in three human HB cell lines (HepT1, HepG2, and HuH6). Following NK1R blockage, we observed decreased phosphorylation of p70S6K and 4E-BP1/2 and inhibition of the canonical Wnt pathway with subsequent decrease of HB cell growth. This effect was dependent of high baseline Wnt activity either by mutational status of β-catenin or extrinsic Wnt activation. Wnt inhibition seemed to be strengthened by disruption of the FOXM1-β-catenin complex. Furthermore, treatment of HB cells with aprepitant led to reduced expression of (liver) stemness markers (AFP, CD13, SOX2, NANOG, and OCT4) and SFA when grown under cancer stem cell conditions. Taken together, we show for the first time that targeting the SP/NK1R signaling cascade inhibits canonical Wnt signaling in HB cells. These findings reveal important insight into the molecular mechanisms of the SP/NK1R complex as a critical component in a model of pediatric liver cancer and may support the development of novel therapeutic interventions for HB and other Wnt-activated cancers.
材料
货号
品牌
产品描述
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
重组人表皮细胞生长因子, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
EGF 人, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
人表皮生长因子,不含动物成分, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
FGF-2 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Sigma-Aldrich
EGF 来源于小鼠, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
Sigma-Aldrich
EGF 来源于大鼠, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
EGF 来源于小鼠, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)