- Widespread expression of human cysteine string proteins.
Widespread expression of human cysteine string proteins.
FEBS letters (1996-08-12)
T Coppola, C Gundersen
PMID8764987
摘要
We present the nucleotide and deduced amino acid sequence of a human systeine string protein (csp) and a unique truncated csp variant that derives from the retention of an exonic sequence that introduces a premature, in-frame stop codon. Low stringency Southern analysis is compatible with the presence of a single human csp gene. Northern analysis reveals that human csp mRNA has both a more heterogeneous size distribution and a more widespread tissue distribution than previously reported for other csps. Exemplifying this is the fact that csp immunoreactivity is detected in Triton X-114 extracts of human blood, an observation which may facilitate blood-based diagnostic assays of csp status in man.