跳转至内容
Merck
  • MiR-146a negatively regulates TLR2-induced inflammatory responses in keratinocytes.
MiR-146a negatively regulates TLR2-induced inflammatory responses in keratinocytes.
The Journal of investigative dermatology (2014-03-29)
Florian Meisgen, Ning Xu Landén, Aoxue Wang, Bence Réthi, Charbel Bouez, Michela Zuccolo, Audrey Gueniche, Mona Ståhle, Enikö Sonkoly, Lionel Breton, Andor Pivarcsi
摘要
Keratinocytes represent the first line of defense against pathogens in the skin and have important roles in initiating and regulating inflammation during infection and autoimmunity. Here we investigated the role of miR-146a in the regulation of the innate immune response of keratinocytes. Toll-like receptor 2 (TLR2) stimulation of primary human keratinocytes resulted in an NF-κB- and mitogen-activated protein kinase-dependent upregulation of miR-146a expression, which was surprisingly long lasting, contrasting with the rapid and transient induction of inflammatory mediators. Overexpression of miR-146a significantly suppressed the production of IL-8, CCL20, and tumor necrosis factor-α, which functionally suppressed the chemotactic attraction of neutrophils by keratinocytes. Inhibition of endogenous miR-146a induced the production of inflammatory mediators even in nonstimulated keratinocytes, and potentiated the effect of TLR2 stimulation. Transcriptomic profiling revealed that miR-146a suppresses the expression of a large number of immune-related genes in keratinocytes. MiR-146a downregulated interleukin-1 receptor-associated kinase 1 and TNF receptor-associated factor 6, two key adapter molecules downstream of TLR signaling, and suppressed NF-κB promoter-binding activity as shown by promoter luciferase experiments. Together, these data identify miR-146a as a regulatory element in keratinocyte innate immunity, which prevents the production of inflammatory mediators under homeostatic conditions and serves as a potent negative feedback regulator after TLR2 stimulation.
材料
货号
品牌
产品描述
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tlr2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Irak1
Sigma-Aldrich
MISSION® esiRNA, targeting human IRAK1