跳转至内容
Merck
  • Increased urinary excretion of the prostaglandin D2 metabolite 9 alpha, 11 beta-prostaglandin F2 after aspirin challenge supports mast cell activation in aspirin-induced airway obstruction.
Increased urinary excretion of the prostaglandin D2 metabolite 9 alpha, 11 beta-prostaglandin F2 after aspirin challenge supports mast cell activation in aspirin-induced airway obstruction.
The Journal of allergy and clinical immunology (1996-08-01)
S O'Sullivan, B Dahlén, S E Dahlén, M Kumlin
摘要
Prostaglandin (PG)D2 is a major product of arachidonic acid metabolism in pulmonary mast cells. We therefore attempted to determine whether measurement of the stable urinary metabolite of PGD2, 9 alpha, 11 beta-PGF2, could serve as a marker of mast cell activation in the lungs. A commercially available enzyme immunoassay was validated and found to be specific and sensitive when applied to unpurified urine. There was no diurnal variation in the levels of 9 alpha, 11 beta-PGF2 in healthy volunteers. Morning baseline values of urinary 9 alpha, 11 beta-PGF2 were measured in three groups--healthy volunteers (n = 9), patients with atopic asthma (n = 14), and aspirin-intolerant patients with asthma (n = 12)--and found to be very similar, 54 +/- 9, 62 +/- 6, and 71 +/- 15 ng/mmol creatinine, respectively (means +/- SEM). Urinary excretion of 9 alpha, 11 beta-PGF2 was increased threefold immediately after allergen-induced bronchoconstriction in nine patients with atopic asthma. Bronchial challenge with inhaled lysine aspirin in eight aspirin-intolerant patients with asthma produced bronchoconstriction without extrapulmonary symptoms and was also followed by a significant increase in the urinary excretion of 9 alpha, 11 beta-PGF2. In addition, challenge with a higher dose of aspirin produced an even greater increase in urinary 9 alpha, 11 beta-PGF2, supporting dose-dependent release of PGD2 during aspirin-induced bronchoconstriction. In contrast, the postchallenge levels of urinary 9 alpha, 11 beta-PGF2 were not increased when bronchoconstriction was induced by histamine challenge in the aspirin-intolerant patients with asthma. The study confirms mast cell involvement in allergen-induced bronchoconstriction and provides novel data, which strongly support the hypothesis that pulmonary mast cells are activated during aspirin-induced airway obstruction. It is finally suggested that measurement of urinary 9 alpha, 11 beta-PGF2 with enzyme immunoassay may be used as a new noninvasive strategy to monitor mast cell activation in vivo.
材料
货号
品牌
产品描述
Sigma-Aldrich
前列腺素 D 2, ≥95%, synthetic