跳转至内容
Merck
Molybdenum cofactor biosynthesis and deficiency.
Cellular and molecular life sciences : CMLS (2005-11-02)
G Schwarz
摘要
The molybdenum cofactor (Moco) forms the active site of all molybdenum (Mo) enzymes, except nitrogenase. Mo enzymes catalyze important redox reactions in global metabolic cycles. Moco consists of Mo covalently bound to one or two dithiolates attached to a unique tricyclic pterin moiety commonly referred to as molybdopterin (MPT). Moco is synthesized by an ancient and conserved biosynthetic pathway that can be divided into four steps, according to the biosynthetic intermediates precursor Z (cyclic pyranopterin monophosphate), MPT and adenylated MPT. In a fifth step modifications such as attachment of nucleotides, sulfuration or bond formation between Mo and the protein result in different catalytic Mo centers. A defect in any of the steps of Moco biosynthesis results in the pleiotropic loss of all Mo enzyme activities. Human Moco deficiency is a hereditary metabolic disorder characterized by severe neurodegeneration resulting in early childhood death. Recently, a first substitution therapy was established.
材料
货号
品牌
产品描述
Sigma-Aldrich
钼, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
钼, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
钼, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
钼, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
钼, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
钼, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
钼, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
钼, wire, diam. 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
钼, foil, thickness 1.0 mm, ≥99.9% trace metals basis