- A simple novel method for studying the combined inhibitory effects of ethylurea and N,N-dimethylurea on jack bean urease.
A simple novel method for studying the combined inhibitory effects of ethylurea and N,N-dimethylurea on jack bean urease.
Journal of enzyme inhibition (1997-01-01)
A A Saboury, A A Moosavi-Movahedi
PMID9204390
摘要
The kinetics of the inhibition of jack bean urease in the presence of ethylurea and N,N-dimethylurea was studied at pH = 7.0. Both inhibitors were competitive inhibitors. A simple novel method was used for determining the dissociation constants (Ki) values for the inhibition which were 26 and 28 mM respectively.
材料