跳转至内容
Merck
  • A simple immunofluorescence technique for simultaneous visualization of mast cells and nerve fibers reveals selectivity and hair cycle--dependent changes in mast cell--nerve fiber contacts in murine skin.
A simple immunofluorescence technique for simultaneous visualization of mast cells and nerve fibers reveals selectivity and hair cycle--dependent changes in mast cell--nerve fiber contacts in murine skin.
Archives of dermatological research (1997-04-01)
V A Botchkarev, S Eichmüller, E M Peters, P Pietsch, O Johansson, M Maurer, R Paus
摘要
Close contacts between mast cells (MC) and nerve fibers have previously been demonstrated in normal and inflamed skin by light and electron microscopy. A key step for any study in MC-nerve interactions in situ is to simultaneously visualize both communication partners, preferably with the option of double labelling the nerve fibers. For this purpose, we developed the following triple-staining technique. After paraformaldehyde-picric acid perfusion fixation, cryostat sections of back skin from C57BL/6 mice were incubated with a primary rat monoclonal antibody to substance P (SP), followed by incubation with a secondary goat-anti-rat TRITC-conjugated IgG. A rabbit antiserum to CGRP was then applied, followed by a secondary goat-anti-rabbit FITC-conjugated IgG. MCs were visualized by incubation with AMCA-labelled avidin, or (for a more convenient quantification of close MC-nerve fiber contacts) with a mixture of TRITC- and FITC-labelled avidins. Using this simple, novel covisualization method, we were able to show that MC-nerve associations in mouse skin are, contrary to previous suggestions, highly selective for nerve fiber types, and that these interactions are regulated in a hair cycle-dependent manner: in telogen and early anagen skin, MCs preferentially contacted CGRP-immunoreactive (IR) or SP/CGRP-IR double-labelled nerve fibers. Compared with telogen values, there was a significant increase in the number of close contacts between MCs and tyrosine hydroxylase-IR fibers during late anagen, and between MCs and peptide histidine-methionine-IR and choline acetyl transferase-IR fibers during catagen.
材料
货号
品牌
产品描述
Sigma-Aldrich
7-氨基-4-甲基香豆素-3-乙酸, BioReagent, suitable for fluorescence, ~90% (HPLC)