- Identification of the catalytic histidine residue participating in the charge-relay system of carboxypeptidase Y.
Identification of the catalytic histidine residue participating in the charge-relay system of carboxypeptidase Y.
Protein science : a publication of the Protein Society (1995-11-01)
G Jung, H Ueno, R Hayashi, T H Liao
PMID8563642
摘要
The essential histidine residue of carboxypeptidase Y (CPY) was modified by a site-specific reagent, a chloromethylketone derivative of benzyloxycarbonyl-L-phenylalanine. The single modified histidine residue was converted to N tau-carboxy-methyl histidine (cmHis) upon performic acid oxidation. A peptide containing cmHis was isolated from the tryptic-thermolytic digest. Based on the amino acid composition and sequence analysis, the peptide is shown to be Val-Phe-Asp-Gly-Gly-cmHis-MetO2-Val-Pro, which was derived from CPY cleaved by trypsin at Arg 391 and thermolysin at Phe 401, and thus His 397 was modified. This histidine residue has been implicated previously by X-ray analysis to participate in the charge-relay system of CPY.