跳转至内容
Merck
  • In vitro toxicity of solubilized 2,3,4-trimethylpentane. I. Cytotoxicity and metabolism of TMP using primary hepatocytes.
In vitro toxicity of solubilized 2,3,4-trimethylpentane. I. Cytotoxicity and metabolism of TMP using primary hepatocytes.
In vitro cellular & developmental biology : journal of the Tissue Culture Association (1989-11-01)
N J Delraso, D R Mattie, C S Godin
摘要
Primary rat hepatocyte suspension cultures (approximately 2 X 10(6) cells) exposed to solubilized 2,3,4-trimethylpentane at concentrations ranging from 7.9 to 31.5 mM under two different culture conditions resulted in a linear dose response, as determined by lactate dehydrogenase leakage and viability data. A significant increase in the 2,3,4-trimethylpentane effective concentration 50 for primary hepatocytes occurred when exposures were implemented in medium containing 0.05% albumin. The effective concentration 50 for hepatocytes exposed to 2,3,4-trimethylpentane in medium lacking and containing albumin were 17.1 and 20.7 mM, respectively. Metabolite analysis by gas chromatography-mass spectrometry of supernatant (lacking or containing albumin) and cell extracts from hepatocyte cultures exposed to 2,3,4-trimethylpentane for 4 h indicated the presence of three metabolites: 2,3,4-trimethyl-1-pentanol, 2,3,4-trimethyl-2-pentanol, 2,3,4-trimethyl-2-pentanol, and 2,3,4-trimethyl-1-pentanoic acid. Electron microscopic examination of 2,3,4-trimethylpentane-exposed primary hepatocytes indicated ultrastructural changes which included abnormal condensed chromatin association with the nuclear membrane, swollen mitochondria, increased amounts of cytoplasmic lipid, significant loss of microvilli from the cell surface, increased vacuolation, and increased numbers of peroxisomes. Although these changes were observed under both culture conditions, they were more severe in cultures lacking albumin. This study indicates that primary hepatocyte suspension cultures provide a useful system for rapidly identifying liver metabolites of selected test compounds of interest.
材料
货号
品牌
产品描述
Sigma-Aldrich
2,3,4-三甲基戊烷, 98%