跳转至内容
Merck
  • Characterization of unique ADTN-catecholamine binding sites in the iris root-ciliary body of rabbits.
Characterization of unique ADTN-catecholamine binding sites in the iris root-ciliary body of rabbits.
Current eye research (1989-12-01)
B H Rohde, G C Chiou
摘要
A binding site for tritiated 2-amino-6, 7-dihydroxy-1, 2,3,4-tetrahydronaphthalene (ADTN) has been partially characterized in the rabbit iris root-ciliary body. Binding of ADTN is proportional to protein content and requires at least 60 minutes to reach equilibrium. Binding is saturable, with a Kd of 27 +/- 1 nM and a Bmax of 2.1 +/- .3 pmol/mg protein (mean +/- SEM). Dopamine competes for this site with a Ki of 100 nM and apomorphine with a Ki of 180 nM. This site is not blocked by L-timolol, phenoxybenzamine, or by several DA1 and DA2 antagonists. It appears to be a new type of catecholamine binding site, of a type not observed outside the anterior eye. It is possible that some of the effects of dopamine on intraocular pressure are mediated through this binding site.