跳转至内容
Merck
  • Opioid peptide deltorphin II simulates the cardioprotective effect of ischemic preconditioning: role of δ₂-opioid receptors, protein kinase C, and K(ATP) channels.
Opioid peptide deltorphin II simulates the cardioprotective effect of ischemic preconditioning: role of δ₂-opioid receptors, protein kinase C, and K(ATP) channels.
Bulletin of experimental biology and medicine (2010-12-18)
L N Maslov, E I Barzakh, A V Krylatov, G A Chernysheva, T Krieg, N V Solenkova, A Yu Lishmanov, S Yu Cybulnikov, Y Zhang
摘要
The cardioprotective properties of a δ₂-opioid receptor agonist deltorphin II were studied in rats with coronary occlusion and reperfusion. Opioid receptor ligands and inhibitors (glybenclamide, chelerythrine, and 5-hydroxydecanoate) were injected intravenously before ischemia and reperfusion. A δ₂-opioid receptor agonist deltorphin II significantly decreased the infarction zone/risk zone index. This effect was abolished by naltrexone, naloxone methiodide, and δ₂-opioid receptor antagonist naltriben, but not by a δ₁-opioid receptor antagonist BNTX. The infarct-limiting effect of deltorphin II was not observed after inhibition of protein kinase C or blockade of mitochondrial K(ATP) channels.
材料
货号
品牌
产品描述
Sigma-Aldrich
[D-Ala2]-Deltorphin II, ≥97% (HPLC)