跳转至内容
Merck
  • Stereoselective metabolism of the environmental mammary carcinogen 6-nitrochrysene to trans-1,2-dihydroxy-1,2-dihydro-6-nitrochrysene by aroclor 1254-treated rat liver microsomes and their comparative mutation profiles in a laci mammary epithelial cell line.
Stereoselective metabolism of the environmental mammary carcinogen 6-nitrochrysene to trans-1,2-dihydroxy-1,2-dihydro-6-nitrochrysene by aroclor 1254-treated rat liver microsomes and their comparative mutation profiles in a laci mammary epithelial cell line.
Chemical research in toxicology (2009-11-06)
Yuan-Wan Sun, Joseph B Guttenplan, Michael Khmelnitsky, Jacek Krzeminski, Telih Boyiri, Shantu Amin, Karam El-Bayoumy
摘要
The environmental pollutant 6-nitrochrysene (6-NC) is a powerful mammary carcinogen and mutagen in rats. Our previous studies have shown that 6-NC is metabolized to trans-1,2-dihydroxy-1,2-dihydro-6-nitrochrysene (1,2-DHD-6-NC) in rats and in several in vitro systems, including human breast tissue, and the latter is the proximate carcinogenic form in the rat mammary gland. Because optically active enantiomers of numerous polynuclear aromatic hydrocarbon (PAH) metabolites including chrysene have different biological activities, we hypothesized that the stereochemical course of 6-NC metabolism might play a significant role in the carcinogenic/mutagenic activities of the parent 6-NC. The goal of this study is to evaluate the effect of stereochemistry on the mutagenicity of 1,2-DHD-6-NC using the cII gene of lacI mammary epithelial cells in vitro. Resolution of (+/-)-1,2-DHD-6-NC was obtained by either nonchiral or chiral stationary phase HPLC methods. We determined that the ratio of (-)-[R,R]- and (+)-[S,S]-1,2-DHD-6-NC formed in the metabolism of 6-NC by rat liver microsomes is 88:12. The mutation fractions and mutation spectra of [R,R] and [S,S]-enantiomers were examined. Our results showed that the [R,R]-isomer is a significantly (p < 0.01) more potent mutagen than the [S,S]-isomer. The major types of mutation induced by the [R,R]-enantiomer are AT > GC, AT > TA, and GC > TA substitutions, and these are similar to those obtained from 6-NC in vivo in the mammary glands of rats treated with 6-NC. The mutation spectra of the [S,S]-isomer were similar to the [R,R]-isomer, but a higher percentage of AT > GC substitutions in the [R,R]-isomer was noted. On the basis of the results of the present study, we hypothesize that [R,R]-1,2-DHD-6-NC is the proximate carcinogen of 6-NC in the rat mammary gland in vivo and will test this hypothesis in a future study.
材料
货号
品牌
产品描述
6-硝基联苯, BCR®, certified reference material