跳转至内容
Merck
  • Medication with antihistamines impairs allergen-specific immunotherapy in mice.
Medication with antihistamines impairs allergen-specific immunotherapy in mice.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology (2007-12-18)
P Johansen, G Senti, J Maria Martínez Gómez, T M Kündig
摘要
Histamine released from activated mast cells and basophils is an important mediator in allergy. Therefore, antihistamines are efficiently and widely used to suppress allergic symptoms. This study evaluated the role of antihistamines in sensitization against allergens and in the efficiency of allergen-specific immunotherapy. CBA mice were sensitized and de-sensitized with bee venom allergen extracts and the major allergen phospholipase A2. Clemastine was used to test the effect of a histamine-1 receptor antagonist on the immune responses to phospholipase A2. The results demonstrated that sensitization against bee venom was strongly enhanced during treatment with antihistamines. Clemastine increased IgE production while decreasing IgG2a production against bee venom. This T-helper type 2 shift of the humoral response appeared to be caused by reduced IFN-gamma and enhanced IL-4 secretion from allergen-specific T cells. We also found reduced TNF-alpha, IL-6 and major histocompatibility complex class-II expression by macrophages. In sensitized mice, the efficiency of allergen-specific immunotherapy was reduced by clemastine treatment. Antihistamines may enhance allergic sensitization and reduce the efficiency of allergen-specific immunotherapy. Future studies will need to demonstrate to what extent pre-medication with antihistamine also affects allergen-specific immunotherapy in humans.
材料
货号
品牌
产品描述
Sigma-Aldrich
克立马丁 富马酸盐, ≥98% (HPLC)