跳转至内容
Merck
  • Absorption spectra of radical forms of 2,4-dihydroxybenzoic acid, a substrate for p-hydroxybenzoate hydroxylase.
Absorption spectra of radical forms of 2,4-dihydroxybenzoic acid, a substrate for p-hydroxybenzoate hydroxylase.
The Journal of biological chemistry (1991-07-15)
R F Anderson, K B Patel, B Vojnovic
摘要
Combined optical and conductimetric measurements in aqueous solution indicate that at high pH (greater than or equal to 10).OH radicals react with the phenoxide form of 2,4-dihydroxybenzoic acid to form transiently phenoxyl radicals and a small amount of hydroxyeyclohexadienyl (HCHD) radicals by 150 ns. The respective yields of 88 and 12% of the total.OH radical yield were deduced from conductance and optical changes as well as from studies using a low potential reductant. The HCHD radical possesses a pKa of 8.0 +/- 0.1 and the constructed spectrum of the deprotonated forms of HCHD has a lambda max at 420 nm with a minimum extinction coefficient of approximately 7250 M-1 cm-1. The red shift in lambda max and increase in extinction coefficient compared to the revised spectral properties of the protonated form of the HCHD radical (lambda max at 390 nm with extinction coefficient of approximately 4500 M-1 cm-1), together with the pKa of the HCHD radical, provide an explanation for the pH-dependent spectral changes of the so-called highly absorbing intermediate II species, observed in the functioning of the enzyme p-hydroxybenzoate hydroxylase. These results add further to the evidence in support of the proposal that intermediate II is composed of species which absorb similarly to the flavin 4(a)-hydroxide and a form of the substrate/product such as the HCHD radical (Anderson, R. F., Patel, K. B., and Stratford, M. R. L. (1987) J. Biol. Chem. 262, 17475-17479).
材料
货号
品牌
产品描述
Sigma-Aldrich
2,4-二羟基苯甲酸, 97%